Skip to Content
Merck
All Photos(1)

Key Documents

45675

Supelco

Terbacil

PESTANAL®, analytical standard

Synonym(s):

3-tert-Butyl-5-chloro-6-methyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13ClN2O2
CAS Number:
Molecular Weight:
216.66
Beilstein:
643054
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1=C(Cl)C(=O)N(C(=O)N1)C(C)(C)C

InChI

1S/C9H13ClN2O2/c1-5-6(10)7(13)12(8(14)11-5)9(2,3)4/h1-4H3,(H,11,14)

InChI key

NBQCNZYJJMBDKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terbacil is a selective and soil applied, uracil herbicide, employed at a pre-emergence stage for the control of annual and some perennial weeds in peaches, citrus and apple orchards. Its mode of action involves the inhibition of photosynthesis process in weeds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Terbacil may be used as an analytical reference standard for the quantification of the analyte in agricultural commodities and fruits using different chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia (2014)
Pesticide multiresidue analysis in cereal grains using modified QuEChERS method combined with automated direct sample introduction GC-TOFMS and UPLC-MS/MS techniques
Mastovska K, et al.
Journal of Agricultural and Food Chemistry, 58(10), 5959-5972 (2009)
Survey for pesticides in wells associated with apple and peach orchards in West Virginia.
H W Hogmire et al.
Bulletin of environmental contamination and toxicology, 44(1), 81-86 (1990-01-01)
M Börzsönyi et al.
Acta morphologica Hungarica, 35(1-2), 3-8 (1987-01-01)
The possible mutagenic effect of the uracil analogue pesticide Terbacil was examined on Drosophila melanogaster and in mammalian cell culture. Terbacil did not prove to be mutagenic in the sex-linked recessive lethal mutation test, and it did not cause an
Multiresidue method for determination of 88 pesticides in berry fruits using solid-phase extraction and gas chromatography-mass spectrometry: Determination of 88 pesticides in berries using SPE and GC-MS.
Yang X, et al.
Food Chemistry, 127(2), 855-865 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service