Skip to Content
Merck
All Photos(1)

Key Documents

444308

Sigma-Aldrich

Tetrabutylammonium bis(3,6-dichloro-1,2-benzenedithiolato)nickelate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H40Cl4NNiS4
CAS Number:
Molecular Weight:
719.39
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

98%

mp

204-206 °C (lit.)

SMILES string

CCCC[N+](CCCC)(CCCC)CCCC.Clc1ccc(Cl)c2S[Ni-]3(Sc12)Sc4c(Cl)ccc(Cl)c4S3

InChI

1S/C16H36N.2C6H4Cl2S2.Ni/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2*7-3-1-2-4(8)6(10)5(3)9;/h5-16H2,1-4H3;2*1-2,9-10H;/q+1;;;+3/p-4

InChI key

DPTWRZYDNJMUTE-UHFFFAOYSA-J


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service