Skip to Content
Merck
All Photos(1)

Key Documents

44344

Supelco

Valeric acid

pharmaceutical impurity standard

Synonym(s):

Valproic acid impurity A (PhEur), n-Valeric acid, Pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COOH
CAS Number:
Molecular Weight:
102.13
Beilstein:
969454
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

vapor density

3.5 (vs air)

Quality Level

vapor pressure

0.15 mmHg ( 20 °C)

API family

valeric acid

Assay

≥99.0% (GC)

autoignition temp.

707 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7.6 %

impurities

≤5.0% water

refractive index

n20/D 1.408 (lit.)

bp

110-111 °C/10 mmHg (lit.)
185 °C (lit.)

mp

−20-−18 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(O)=O

InChI

1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)

InChI key

NQPDZGIKBAWPEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service