Skip to Content
Merck
All Photos(1)

Key Documents

43825

Supelco

Salicylic acid (phenyl-13C6)

analytical standard

Synonym(s):

2-Hydroxybenzoic acid (phenyl-13C6)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€1,150.00

€1,150.00


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
€1,150.00

About This Item

Linear Formula:
2-(HO)13C6H4CO2H
CAS Number:
Molecular Weight:
144.08
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€1,150.00


Available to ship on01 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

158-161 °C (lit.)

application(s)

food and beverages
forensics and toxicology
pharmaceutical
veterinary

format

neat

mass shift

M+6

storage temp.

2-8°C

SMILES string

OC(=O)[13c]1[13cH][13cH][13cH][13cH][13c]1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

YGSDEFSMJLZEOE-IDEBNGHGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

LC-MS identity conforms

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service