Skip to Content
Merck
All Photos(1)

Key Documents

43612

Supelco

Glyoxal solution

~40% in H2O, for HPLC derivatization, LiChropur

Synonym(s):

Ethanedial, Oxalaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHCCHO
CAS Number:
Molecular Weight:
58.04
Beilstein:
1732463
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

form

liquid

quality

LiChropur

concentration

38.0-42.0%
~40% in H2O

technique(s)

HPLC: suitable

SMILES string

[H]C(=O)C([H])=O

InChI

1S/C2H2O2/c3-1-2-4/h1-2H

InChI key

LEQAOMBKQFMDFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable for HPLC derivatization

Other Notes

This form of glyoxal is composed of 3 moles of glyoxal and 2 moles of water in a relatively stable configuration.

Caution

May precipitate on storage; redissolve at 50-60 °C.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precolumn derivatization with glyoxal as a new approach to the highly sensitive HPLC-UV determination of unsymmetrical dimethylhydrazine.
Smirnov, R. S., et al.
Journal of Analytical Chemistry, 68, 837-844 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service