Skip to Content
Merck
All Photos(1)

Key Documents

43265

Millipore

Antibiotic agar No. 5

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€247.00

€247.00


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
€247.00

About This Item

UNSPSC Code:
41171608
NACRES:
NA.74

€247.00


Available to ship on02 April 2025Details


Request a Bulk Order

form

solid

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
beef extract, 1.5 g/L
peptone, 6.0 g/L
yeast extract, 3.0 g/L

final pH

7.9±0.2 (25 °C)

application(s)

microbiology

Application

Antibiotic Agar No.5 is used for microbiological assay of Dihydrostreptomycin, Framycetin and Kanamycin B using Bacillus subtilis.

Preparation Note

Suspend 25.50 grams in 1000 ml purified /distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool to 45-50°C. Mix well and pour into sterile Petri plates.
Advice: Recommended for the Microbiological assay of Dihydrostreptomycin, Framycetin, Kanamycin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service