Skip to Content
Merck
All Photos(1)

Documents

42641

Supelco

(Z)-3-Methylglutaconic acid

≥97.0% (HPLC)

Synonym(s):

(2Z)-3-Methyl-2-pentenedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
Beilstein:
1722908
UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

Assay

≥97.0% (HPLC)

mp

147-151 °C

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C/C(CC(O)=O)=C/C(O)=O

InChI

1S/C6H8O4/c1-4(2-5(7)8)3-6(9)10/h2H,3H2,1H3,(H,7,8)(H,9,10)/b4-2+

InChI key

WKRBKYFIJPGYQC-DUXPYHPUSA-N

Biochem/physiol Actions

2-Methylglutaconic acid is found in the urine of patients with organic aciduria from 2-methylacetoacetyl-CoA thiolase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian J Wong et al.
Biochemistry, 43(16), 4646-4654 (2004-04-21)
Members of the enoyl-CoA hydratase (crotonase) superfamily catalyze different overall reactions that utilize a common catalytic strategy delivered by a shared structural scaffold; the substrates are usually acyl esters of coenzyme A, and the intermediates are usually thioester enolate anions
Utilization of branched chain acids in cholesterol synthesis.
K BLOCH et al.
The Journal of biological chemistry, 211(2), 687-699 (1954-12-01)
W Meier-Augenstein et al.
Journal of chromatography, 615(1), 127-135 (1993-05-19)
An improved method for the identification and quantification of organic acids in body fluids employing capillary gas chromatography-mass spectrometry has been developed. A thick-film capillary column, that combines the properties of a capillary column with those of a megabore column
Rebecca Young et al.
Biochemical and biophysical research communications, 534, 261-265 (2020-12-08)
3-methylglutaconic (3MGC) aciduria is associated with a growing number of discrete inborn errors of metabolism. Herein, an antibody-based approach to detection/quantitation of 3MGC acid has been pursued. When trans-3MGC acid conjugated keyhole limpet hemocyanin (KLH) was inoculated into rabbits a
M Duran et al.
Biomedical mass spectrometry, 9(1), 1-5 (1982-01-01)
The identification of (E)-2-methylglutaconic acid, a 'new' metabolite of isoleucine, is described. The substance was detected in urine samples from patients with propionic acidaemia, methylmalonic acidaemia and so-called beta-ketothiolase deficiency; in the majority of cases together with N-tiglylglycine. (E)-2-Methylglutaconic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service