Skip to Content
Merck
All Photos(1)

Key Documents

424455

Sigma-Aldrich

Resorufin

Dye content 95 %

Synonym(s):

7-Hydroxy-3H-phenoxazin-3-one, NSC 12097

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7NO3
CAS Number:
Molecular Weight:
213.19
Beilstein:
174850
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

form

powder

Quality Level

composition

Dye content, 95%

mp

>300 °C (lit.)

λmax

573 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O=C(C=C1)C=C2C1=NC3=CC=C(O)C=C3O2

InChI

1S/C12H7NO3/c14-7-1-3-9-11(5-7)16-12-6-8(15)2-4-10(12)13-9/h1-6,14H

InChI key

HSSLDCABUXLXKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Resorufin, a redox probe can be used to visualize cell respiration directly.[1] It is highly fluorescent and has an excitation maximum at 563nm and emission maximum at 587nm.[2]

Application

Resorufin has been used in the measurements of hydrogen peroxide generation.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yutaka Hitomi et al.
Analytical chemistry, 83(24), 9213-9216 (2011-11-18)
We developed a metal-based fluorescent probe for H(2)O(2) called MBFh1, which has an iron complex as a reaction site for H(2)O(2) and a 3,7-dihydroxyphenoxazine derivative as the fluorescent reporter unit. The iron complex reacts quickly with H(2)O(2) to form oxidants
Hubert M Piwonski et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(22), E1437-E1443 (2012-05-09)
Enzymatic inhibition by product molecules is an important and widespread phenomenon. We describe an approach to study product inhibition at the single-molecule level. Individual HRP molecules are trapped within surface-tethered lipid vesicles, and their reaction with a fluorogenic substrate is
Eyal Golub et al.
The Analyst, 136(21), 4397-4401 (2011-09-02)
Hemin/G-quadruplex catalyzes the H(2)O(2)-mediated oxidation of Amplex Red to the fluorescent product resorufin. This process is implemented to develop hairpin nucleic acid structures for the detection of DNA, to probe the catalytic activity of glucose oxidase, to use the thrombin-aptamer
Malena B Rone et al.
Molecular endocrinology (Baltimore, Md.), 26(11), 1868-1882 (2012-09-14)
Steroid hormones are critical for organismal development and health. The rate-limiting step in steroidogenesis is the transport of cholesterol from the outer mitochondrial membrane (OMM) to the cytochrome P450 enzyme CYP11A1 in the inner mitochondrial membrane (IMM). Cholesterol transfer occurs
Eduardo Vottero et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 16(6), 899-912 (2011-05-14)
CYP102A1, originating from Bacillus megaterium, is a highly active enzyme which has attracted much attention because of its potential applicability as a biocatalyst for oxidative reactions. Previously we developed drug-metabolizing mutant CYP102A1 M11 by a combination of site-directed and random

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service