Skip to Content
Merck
All Photos(1)

Key Documents

41513

Supelco

2-Hexanol

analytical standard

Synonym(s):

(±)-2-Hexanol, Butyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718996
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.414 (lit.)
n20/D 1.415

bp

136 °C (lit.)

density

0.814 g/mL at 20 °C
0.81 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCC(C)O

InChI

1S/C6H14O/c1-3-4-5-6(2)7/h6-7H,3-5H2,1-2H3

InChI key

QNVRIHYSUZMSGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hexanol is a volatile aromatic alcohol abundantly occurring in various plant materials. It is often found as a constituent of food and beverages.[1][2][3][4]

Application

2-Hexanol may be used as an analytical standard for the determination of the analyte in biological samples,[5][6][7] fruits,[1] soymilk,[2] food,[3] and aqueous samples[8] by various chromatography techniques.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of volatile compounds in soymilk using solid-phase microextraction-gas chromatography
Achouri A, et al.
Food Chemistry, 99(4), 759-766 (2006)
F Schade et al.
Phytochemistry, 56(7), 703-710 (2001-04-21)
Thirteen major volatiles of the carnation flower fragrance signature have been identified by GC/MS. Of these, ten, hexanal, (2E)-hexenal, 1-hexanol, 2-hexanol, 3-hexen-1-ol, nonanal, benzaldehyde, benzyl alcohol, benzyl benzoate and caryophyllene, were quantified. The steady-state levels of these ten volatiles change
Analysis of volatile flavor components in roasted peanuts using supercritical fluid extraction and gas chromatography- mass spectrometry
Leunissen M, et al.
Journal of Agricultural and Food Chemistry, 44(9), 2694-2699 (1996)
Analysis of n-hexane, 2-hexanone, 2, 5-hexanedione, and related chemicals by capillary gas chromatography and high-performance liquid chromatography
Nomeir AA and Abou-Donia MB
Analytical biochemistry, 151(2), 381-388 (1985)
L Perbellini et al.
International archives of occupational and environmental health, 48(1), 99-106 (1981-01-01)
A gas chromatographic method for analyzing the urinary metabolites of n-hexane (2-hexanol, 2,5-hexanedione, 2,5-dimethylfuran and gamma-valerolactone), of 2-methylpentane (2-methyl-2-pentanol), of 3-methylpentane (3-methyl-2-pentanol), and of cyclohexane (cyclohexanol) was developed. Processing of urine and the gas chromatographic conditions are described. The recovery

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service