Skip to Content
Merck
All Photos(1)

Key Documents

39411

Supelco

Polystyrene Beads for Gel Permeation Chromatography

for organic compounds with molecular weight up to 1000 daltons

Synonym(s):

Gel Permeation Chromatography Polystyrene Beads, Size exclusion Chromatography Polystyrene Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
SB.52

Quality Level

form

beads

technique(s)

LPLC: suitable

matrix

crosslinked with 8% DVB

matrix active group

polymer

particle size

40-80 μm

separation technique

size exclusion (SEC)

General description

Polystyrene (PS) is a widely used polymer due to properties such as low specific weight, high chemical resistance and mechanical flexibility and also it is biocompatible. PS as biomaterials was manufactured for cell attachment. Functionalized porous polymer beads are used for solid-phase organic synthesis, combinatorial chemistry, polymer-supported catalysis, ion-exchange resin. Polystyrene gels are cross-linked in presence of diluents which forms a fine-mesh bead used for packing into chromatographic columns.

Application

Polystyrene Beads may be used as packing material for columns used in Gel Permeation Chromatography.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gel permeation chromatography. I. A new method for molecular weight distribution of high polymers.
Moore, J. C.
Journal of Polymer Science: Part A, General Papers, General papers 2, 835-843 (1964)
Surface modification of polystyrene beads by ultraviolet/ozone treatment and its effect on gelatin coating.
Yusilawati, A. N., et al.
American Journal of Applied Sciences , 7, 724-724 (2010)
Plasmachemical amine functionalization of porous polystyrene beads: the importance of pore architecture.
?ye, G., et al.
Langmuir, 18, 8996-8999 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service