Skip to Content
Merck
All Photos(1)

Documents

38763

Sigma-Aldrich

4′,7-Dimethoxyisoflavone

≥96.0% (TLC)

Synonym(s):

Daidzein dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O4
CAS Number:
Molecular Weight:
282.29
Beilstein:
265550
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥96.0% (TLC)

SMILES string

COc1ccc(cc1)C2=COc3cc(OC)ccc3C2=O

Biochem/physiol Actions

Plant isoflavone that promotes proliferation of mitochondria in cells damaged by toxins, trauma, or hypoxia. Activity is mediated through the activation and expression of sirtuin (SIRT1), a deacetylase and activator of PGC-1α.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service