Skip to Content
Merck
All Photos(1)

Key Documents

37305

Sigma-Aldrich

3-Hydroxy-1,2,3-benzotriazin-4(3H)-one

purum, ≥98.0% (T)

Synonym(s):

3,4-Dihydro-3-hydroxy-4-oxo-1,2,3-benzotriazine, 3-Hydroxy-3,4-dihydro-4-oxo-1,2,3-benzotriazine, DHBT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
Beilstein:
609382
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Assay

≥98.0% (T)

mp

~180 °C (dec.)

SMILES string

ON1N=Nc2ccccc2C1=O

Looking for similar products? Visit Product Comparison Guide

Other Notes

Additive which suppresses racemization in peptide synthesis; Synthesis of amino acid active esters used as self-indicating building blocks in solid-phase synthesis

Pictograms

Exploding BombExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Expl. 1.1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

1 - Explosive Substances

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.R. Cameron et al.
Journal of the Chemical Society. Perkin Transactions 1, 2895-2895 (1988)
[Racemization in peptide syntheses].
W König et al.
Chemische Berichte, 103(7), 2024-2033 (1970-07-01)
Y. Kurano et al.
Journal of the Chemical Society. Chemical Communications, 323-323 (1987)
E. Atherton et al.
Journal of the Chemical Society. Perkin Transactions 1, 2887-2887 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service