Skip to Content
Merck
All Photos(2)

Key Documents

37007

Supelco

Iminoctadine triacetate

PESTANAL®, analytical standard

Synonym(s):

N,N″′-(Iminodi-8,1-octanediyl)bisguanidine triacetate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€379.00

€379.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
10 MG
€379.00

About This Item

Empirical Formula (Hill Notation):
C24H53N7O6
CAS Number:
Molecular Weight:
535.72
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€379.00


In StockDetails


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(O)=O.CC(O)=O.CC(O)=O.NC(=N)NCCCCCCCCNCCCCCCCCNC(N)=N

InChI

1S/C18H41N7.3C2H4O2/c19-17(20)24-15-11-7-3-1-5-9-13-23-14-10-6-2-4-8-12-16-25-18(21)22;3*1-2(3)4/h23H,1-16H2,(H4,19,20,24)(H4,21,22,25);3*1H3,(H,3,4)

InChI key

FKWDSATZSMJRLC-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinbong Park et al.
Frontiers in pharmacology, 10, 1458-1458 (2020-01-11)
Atopic dermatitis (AD) is an inflammatory disease of the skin, resulting from an immune dysfunction, that often occurs as a comorbidity of obesity. This investigation evaluated the capacity of Taeumjowi-tang (TJT), a Korean herbal formulation from the Sasang medical tradition
Jihye Min et al.
Medicine, 96(32), e7737-e7737 (2017-08-11)
The purpose of this study was to identify the chemical responsible for cholestatic hepatitis in a 55-year-old woman who ingested 1,1'-iminodi (octamethylene) diguanidinium triacetate (iminoctadine triacetate), a fungicide. The fungicide formulation was also composed of polyoxyethylene nonylphenol (NP-40) and methanol.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service