Skip to Content
Merck
All Photos(1)

Key Documents

36667

Supelco

Atrazine-desethyl-desisopropyl

PESTANAL®, analytical standard

Synonym(s):

6-Chloro-2,4-diamino-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4ClN5
CAS Number:
Molecular Weight:
145.55
Beilstein:
124340
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1nc(N)nc(Cl)n1

InChI

1S/C3H4ClN5/c4-1-7-2(5)9-3(6)8-1/h(H4,5,6,7,8,9)

InChI key

FVFVNNKYKYZTJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Atrazine-desethyl-desisopropyl may be used as an analytical reference standard for the quantification of the analyte in environmental samples using different chromatography techniques
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On-line solid-phase extraction method for determination of triazine herbicides and degradation products in seawater by ultra-pressure liquid chromatography--tandem mass spectrometry
Rodriguez-Gonzalez.N, et al.
Journal of Chromatography A, 1470(2), 33-41 (2016)
Determination of 48 pesticides and their main metabolites in water samples by employing sonication and liquid chromatography--tandem mass spectrometry
Fenoll J, et al.
Talanta, 85(2), 975-982 (2011)
Joseane M Pozzebon et al.
Journal of chromatography. A, 987(1-2), 375-380 (2003-03-05)
A method was developed to determine simazine, atrazine and their metabolite, 2-chloro-4,6-diamino-1,3,5-triazine, in urine. The presence of these herbicides in urine may reflect possible exposure to pesticides. Sample preparation involved protein precipitation and solid-phase extraction. The samples were analyzed by
Greg P Dooley et al.
Chemical research in toxicology, 19(5), 692-700 (2006-05-16)
Atrazine (2-chloro-4-[ethylamino]-6-[isopropylamino]-1,3,5-triazine) is one of the most commonly used herbicides in North America and is frequently detected in ground and surface waters. This research investigated possible covalent modifications of hemoglobin following in vivo exposures to atrazine in Sprague Dawley (SD)
Jennifer L Seffernick et al.
Applied and environmental microbiology, 68(9), 4672-4675 (2002-08-30)
2-Chloro-4,6-diamino-s-triazine (CAAT) is a metabolite of atrazine biodegradation in soils. Atrazine chlorohydrolase (AtzA) catalyzes the dechlorination of atrazine but is unreactive with CAAT. In this study, melamine deaminase (TriA), which is 98% identical to AtzA, catalyzed deamination of CAAT to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service