Skip to Content
Merck
All Photos(1)

Key Documents

34555

Supelco

2-(Thiocyanatomethylthio)benzothiazole

PESTANAL®, analytical standard

Synonym(s):

(1,3-Benzothiazol-2-ylthio)methyl thiocyanate, 2-(Thiocyanatomethylthio)-1,3-benzothiazole, Benthiazole, TCMTB, Thiocyanic acid 2-(benzothiazolylthio)methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2S3
CAS Number:
Molecular Weight:
238.35
Beilstein:
1213014
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

SMILES string

N#CSCSC1=NC(C=CC=C2)=C2S1

InChI

1S/C9H6N2S3/c10-5-12-6-13-9-11-7-3-1-2-4-8(7)14-9/h1-4H,6H2

InChI key

TUBQDCKAWGHZPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela R Buckalew et al.
Archives of toxicology, 94(3), 873-885 (2020-02-18)
The Fischer rat thyroid follicular cell line (FRTL-5) endogenously expresses the sodium-iodide symporter (NIS) and has been used to identify environmental chemicals that perturb thyroid hormone homeostasis by disruption of NIS-mediated iodide uptake. Previously, a high-throughput radioactive iodide uptake (RAIU)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service