Skip to Content
Merck
All Photos(2)

Key Documents

34324

Supelco

Coumafuryl

PESTANAL®, analytical standard

Synonym(s):

3-(α-Acetonyl-2-furylmethyl)-4-hydroxycoumarin, 3-(α-Acetonylfuryl)-4-hydroxy-coumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O5
CAS Number:
Molecular Weight:
298.29
Beilstein:
6817711
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(=O)CC(c1ccco1)C2=C(O)c3ccccc3OC2=O

InChI

1S/C17H14O5/c1-10(18)9-12(13-7-4-8-21-13)15-16(19)11-5-2-3-6-14(11)22-17(15)20/h2-8,12,19H,9H2,1H3

InChI key

JFIXKFSJCQNGEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 3 - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>413.6 °F - closed cup

Flash Point(C)

> 212 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L L Munger et al.
Avian diseases, 37(2), 622-624 (1993-04-01)
Coumafuryl (Fumarin) toxicity was diagnosed in chickens less than 1 week of age. Mortality rate was 100%. Necropsy showed crops and gizzards to be full of feed. There was diffuse hemorrhage and unclotted blood in the abdomen and thorax. Histological
Yukari Nakamura et al.
Journal of chromatography. A, 1516, 71-78 (2017-08-19)
Monodisperse molecularly imprinted polymers (MIPs) for warfarin (WF) and coumachlor (CC), MIP
D M Renapurkar
Journal of hygiene, epidemiology, microbiology, and immunology, 26(2), 125-130 (1982-01-01)
Results are presented of feeding tests carried out with three common anticoagulant rodenticides viz., coumatetralyl, fumarin and warfarin on three common species of commensal rodents i.e., Rattus rattus, Rattus norvegicus and Bandicota bengalensis. All three species of rodents were susceptible
Zheng Qiao et al.
Journal of forensic sciences, 63(3), 784-792 (2018-05-04)
Anticoagulant rodenticides are widely used for rodent control around the world. A rapid and sensitive method was developed and validated for the simultaneous determination of 13 anticoagulant rodenticides (coumafuryl, pindone, valone, warfarin, coumatetralyl, coumachlor, diphacinone, dicumarol, chlorophacinone, bromadiolone, difenacoum, flocoumafen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service