Skip to Content
Merck
All Photos(1)

Key Documents

34321

Supelco

Trichloronitromethane

PESTANAL®, analytical standard

Synonym(s):

Chloropicrin, Nitrotrichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CNO2
CAS Number:
Molecular Weight:
164.38
Beilstein:
1756135
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.461 (lit.)

bp

112 °C (lit.)

density

1.657 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

[O-][N+](=O)C(Cl)(Cl)Cl

InChI

1S/CCl3NO2/c2-1(3,4)5(6)7

InChI key

LFHISGNCFUNFFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianrong Wei et al.
The Science of the total environment, 408(20), 4600-4606 (2010-07-29)
Disinfection by-products were determined in 15 water treatment plants in Beijing City. The effects of different water sources (surface water source, mixture water source and ground water source), seasonal variation and spatial variation were examined. Trihalomethanes and haloacetic acids were
D Wang et al.
Journal of environmental quality, 40(5), 1443-1449 (2011-08-27)
Understanding the control mechanisms of fumigant movement in soil is a fundamental step for developing management strategies to reduce atmospheric emissions. Most soil fumigants including chloropicrin (CP) are applied by shank injection, and the application process often leaves vertical soil
M Pesonen et al.
Toxicology letters, 211(3), 239-245 (2012-04-21)
Chloropicrin is an aliphatic volatile nitrate compound that is mainly used as a pesticide. It has several toxic effects in animals and can cause irritating and other health problems in exposed humans. Since the mode of chloropicrin action is poorly
Qiuxia Wang et al.
Journal of environmental quality, 39(3), 917-922 (2010-04-20)
Chloropicrin (CP) is highly volatile and may pose strong exposure risks for humans and the environment. A gelatin capsule formulation was developed to reduce atmospheric CP emissions and to improve application safety. The objective of this study was to determine
Dongdong Yan et al.
Journal of agricultural and food chemistry, 60(20), 5023-5027 (2012-05-04)
Gelatin capsules containing chloropicrin (Pic gel cap) were developed as a new formulation to reduce the potential human exposure risks associated with injection application methods. The objective of this study was to test the efficacy of a Pic gel cap

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service