Skip to Content
Merck
All Photos(1)

Key Documents

34301

Supelco

Dimethyltin dichloride

PESTANAL®, analytical standard

Synonym(s):

Dichlorodimethylstannane, Dichlorodimethyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SnCl2
CAS Number:
Molecular Weight:
219.69
Beilstein:
4123349
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

101-106 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

C[Sn](C)(Cl)Cl

InChI

1S/2CH3.2ClH.Sn/h2*1H3;2*1H;/q;;;;+2/p-2

InChI key

PKKGKUDPKRTKLJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1

Target Organs

Immune system,Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service