Skip to Content
Merck
All Photos(2)

Key Documents

32918

Supelco

Halosulfuron-methyl

PESTANAL®, analytical standard

Synonym(s):

Methyl 3-chloro-5-(4,6-dimethoxy-2-pyrimidinylcarbamoylsulfamoyl)-1-methylpyrazole-4-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€113.00

€113.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 MG
€113.00

About This Item

Empirical Formula (Hill Notation):
C13H15ClN6O7S
CAS Number:
Molecular Weight:
434.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€113.00


Available to ship on07 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1c(Cl)nn(C)c1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2

InChI

1S/C13H15ClN6O7S/c1-20-10(8(9(14)18-20)11(21)27-4)28(23,24)19-13(22)17-12-15-6(25-2)5-7(16-12)26-3/h5H,1-4H3,(H2,15,16,17,19,22)

InChI key

FMGZEUWROYGLAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Halosulfuron-methyl is a highly active postemergence sulfonylurea herbicide, which finds applications in weed control management.[1][2]

Application

Halosulfuron-methyl may be used as a standard in the determination of halosulfuron methyl in water samples using liquid chromatography coupled with electrospray ionization mass spectrometry (LC/ES-MS).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformation kinetics and mechanism of the sulfonylurea herbicides pyrazosulfuron ethyl and halosulfuron methyl in aqueous solutions.
Zheng W, et al.
Journal of Agricultural and Food Chemistry, 56(16), 7367-7372 (2008)
Confirmation and quantitation of selected sulfonylurea, imidazolinone, and sulfonamide herbicides in surface water using electrospray LC/MS.
Rodriguez M and Orescan B D.
Analytical Chemistry, 70(13), 2710-2717 (1998)
Determination and quantitation of sulfonylurea and urea herbicides in water samples using liquid chromatography with electrospray ionization mass spectrometric detection.
Ayano E, et al.
Analytica Chimica Acta, 507(2), 211-218 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service