Skip to Content
Merck
All Photos(1)

Documents

31642-M

Sigma-Aldrich

Hydrogen peroxide solution

30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.

Synonym(s):

Hydrogen peroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
NACRES:
NA.21

product name

Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Quality Level

vapor pressure

23.3 mmHg ( 30 °C)

grade

puriss. p.a.

Assay

30% (w/w)

form

liquid

quality

stabilized

shelf life

Shelf life is limited.

impurities

≤0.005% non-volatile matter
≤30 mg/kg total nitrogen (N)
0.5-5.0 mEq/L titrable acid (as H2SO4)

ign. residue

≤0.002%

density

1.11 g/cm3

anion traces

chloride (Cl-): ≤0.5 mg/kg
phosphate (PO43-): ≤30 mg/kg
sulfate (SO42-): ≤1 mg/kg

cation traces

Al: ≤0.00005%
As: ≤0.00005%
Ba: ≤0.000005%
Be: ≤0.000001%
Bi: ≤0.00001%
Ca: ≤0.00002%
Cd: ≤0.000001%
Co: ≤0.000001%
Cr: ≤0.000002%
Cu: ≤0.000001%
Fe: ≤0.000005%
Ge: ≤0.000005%
K: ≤0.0005%
Li: ≤0.000001%
Mg: ≤0.000005%
Mn: ≤0.000001%
Mo: ≤0.000002%
Na: ≤0.0005%
Ni: ≤0.000002%
Pb: ≤0.000001%
Sr: ≤0.000001%
Ti: ≤0.00001%
Tl: ≤0.000005%
V: ≤0.000001%
Zn: ≤0.000005%
Zr: ≤0.00001%

SMILES string

OO

storage temp.

2-8°C

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This 30% H2O2 (w/w) is a solution of hydrogen peroxide. Its decomposition in the presence of varying concentrations of zeolites and manganese (catalyst) has been investigated.

Application

This 30% H2O2 (w/w) was employed as a catalyst in a chemical treatment method for industrial waste water contaminated with unconventional oil (oil shale).

Caution

keep in dark area

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service