Skip to Content
Merck
All Photos(1)

Key Documents

29920

Sigma-Aldrich

Diethyl cyclopropane-1,1-dicarboxylate

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H4(CO2C2H5)2
CAS Number:
Molecular Weight:
186.21
Beilstein:
2049509
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

contains

~0.05% hydroquinone monomethyl ether as stabilizer

refractive index

n20/D 1.433 (lit.)
n20/D 1.433

bp

94-96 °C/10 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1(CC1)C(=O)OCC

InChI

1S/C9H14O4/c1-3-12-7(10)9(5-6-9)8(11)13-4-2/h3-6H2,1-2H3

InChI key

KYYUCZOHNYSLFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl cyclopropane-1,1-dicarboxylate undergoes ring-opening addition reactions with various nucleophilic reagents.

Other Notes

Review of electrophilic cyclopropanes

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Danishefsky
Accounts of Chemical Research, 12, 66-66 (1979)
Nucleophilic ring-opening additions to 1, 1-disubstituted cyclopropanes.
Stewart JM and Westberg HH.
The Journal of Organic Chemistry, 30(6), 1951-1955 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service