Skip to Content
Merck
All Photos(1)

Key Documents

28976

Millipore

Thioglycollate Medium with K Agar

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

solid

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
dextrin, 2.75 g/L
maltose, 12.75 g/L
peptic digest of animal tissue, 0.78 g/L

final pH

4.7±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

Micrococcus spp.
nonselective for Bacillus spp.
nonselective for Bacteroides spp.
nonselective for Candida spp.
nonselective for Clostridium spp.
nonselective for Neisseria spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for molds (General Media)
nonselective for yeasts (General Media)

Application

Recommended for isolation, detection and enumeration of yeasts and molds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service