28651-U
Hamilton® 1710N syringe
1710N Slim Line, volume 100 μL, needle size 26s ga (cone tip), needle L 51 mm (2 in.)
About This Item
Recommended Products
material
cone tip stainless steel (point style 1)
needle size
26s ga (cone tip)
description
gastight
feature
GASTIGHT® Syringe, Fixed Needle
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 203076
needle L
51 mm (2 in.)
volume
100 μL
compatibility
for use with CTC A200S PAL Instruments (cemented needle)
Looking for similar products? Visit Product Comparison Guide
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service