Skip to Content
Merck
All Photos(1)

Documents

17156

Millipore

ITC Broth (Base)

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Irgasan Ticarcillin Chlorate Broth (Base), TTC Broth (Base), Triclosan Ticarcillin Chlorate Broth (Base)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

Agency

according to ISO 10273:2017

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein enzymic hydrolysate, 10 g/L
irgasan (trichlosan), 0.001 g/L
magnesium chloride.6H2O, 60 g/L
malachite green, 0.01 g/L
sodium chloride, 5 g/L
yeast extract, 1 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.9±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages

microbiology

suitability

selective for Yersinia spp.

Application

For selective enrichment, enumeration of Yersinia enterocolitica.

Preparation Note

Suspend 76 g in 970 ml distilled water. Heat to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes. Cool to 45-50°C. Aseptically add 1 vial of each Ticarcillin Supplement (Cat. No. 17778) and Potassium Chlorate Supplement (Cat. No. 17777). Mix well before dispensing in sterile tubes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service