Skip to Content
Merck
All Photos(3)

Documents

126225

Sigma-Aldrich

Chromotropic acid disodium salt dihydrate

technical grade

Synonym(s):

1,8-Dihydroxynaphthalene-3,6-disulfonic acid disodium salt, 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C10H4(SO3Na)2·2H2O
CAS Number:
Molecular Weight:
400.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

technical grade

Quality Level

Assay

≥78.5%

form

powder

mp

>300 °C (lit.)

SMILES string

O.O.[Na+].[Na+].Oc1cc(cc2cc(cc(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C10H8O8S2.2Na.2H2O/c11-8-3-6(19(13,14)15)1-5-2-7(20(16,17)18)4-9(12)10(5)8;;;;/h1-4,11-12H,(H,13,14,15)(H,16,17,18);;;2*1H2/q;2*+1;;/p-2

InChI key

QUEAKWJKJBFNEG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chromotropic acid disodium salt dihydrate is a tan colored sodium salt.

Application

Chromotropic acid disodium salt dihydrate may be employed as a chromogenic reagent for the quantification of dipyrone in various pharmaceutical preparations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Spectrophotometric determination of dipyrone in pharmaceutical preparations by using chromotropic acid"
Sakiara KA, et al.
Il Farmaco; Edizione Scientifica, 54(09), 629-635 (1999)
Spot-test identification and rapid quantitative sequential analysis of dipyrone.
Tubino M, et al.
Ecletica Quimica , 35(10), 41-46 (2010)
Rapid Spot Test Analysis for the Detection of Dipyrone in Pharmaceutical Preparations.
Pezza L, et al.
Analytical Sciences, 16(3), 313-315 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service