Skip to Content
Merck
All Photos(1)

Key Documents

12448

Supelco

α-Humulene

analytical standard

Synonym(s):

α-Caryophyllene, trans,trans,trans-2,6,6,9-Tetramethyl-1,4,8-cycloundecatriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
3240075
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.503 (lit.)

bp

166-168 °C (lit.)

density

0.889 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C\C1=C/CC(C)(C)\C=C\C\C(C)=C\CC1

InChI

1S/C15H24/c1-13-7-5-8-14(2)10-12-15(3,4)11-6-9-13/h6-7,10-11H,5,8-9,12H2,1-4H3/b11-6+,13-7+,14-10+

InChI key

FAMPSKZZVDUYOS-HRGUGZIWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Humulene is a cyclic sesquiterpene, a main active constituent isolated from plant Cordia verbenacea (Boraginaceae).

Application

α-Humulene has been used as a reference standard for the determination of isomers of α-humulene in copaiba oleoresin species by high performance liquid chromatography method using the Box-Behnken design.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Where is the Oxygen? Structural Analysis of ?-Humulene Oxidation Products by the Crystalline Sponge Method.
Zigon N, et al.
Angewandte Chemie (International Edition in English), 54(31), 9033-9037 (2015)
Qiu-Yue Qi et al.
Organic letters, 16(19), 5092-5095 (2014-09-13)
Two new heterodimeric sesquiterpenes, sterhirsutins A (1) and B (2), and two new sesquiterpenes, hirsutic acids D-E (3 and 4), were identified from the culture of Stereum hirsutum. The absolute configurations in 1 and 2 were confirmed by single-crystal X-ray
Development of a high performance liquid chromatography method for quantification of isomers β-caryophyllene and α-humulene in copaiba oleoresin using the Box-Behnken design.
de Almeida Borges VR, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 940(14), 35-41 (2013)
Juliana Siqueira Chaves et al.
Planta medica, 74(14), 1678-1683 (2008-10-28)
A quantitative study was undertaken to assess the plasma and tissue levels, tissue distribution and skin (ear) absorption of the sesquiterpene alpha-humulene, the main active constituent isolated from the plant Cordia verbenacea (Boraginaceae ), after oral, intravenous and topical administration

Protocols

GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service