Skip to Content
Merck
All Photos(1)

Key Documents

12242

Supelco

Tin-base solder (lead free)

manufactured by MBH Analytical Ltd (74X AM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

material

disks

grade

analytical standard

technique(s)

XRF: suitable

diam. × H

40 mm × 15 mm , dimensions are approximate

format

matrix material

Analysis Note

Values of analytes vary lot to lot.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactive diffusion between Ag and Sn at solid state temperatures.
Suzuki, Ken, et al.
Materials Transactions, 46.5, 969-973 (2005)
Flux development for lead-free solders containing zinc.
Vaynman, S., and M. E. Fine.
Journal of Electronic Materials, 29.10, 1160-1163 (2000)
Nan Song et al.
Molecular cell, 68(1), 185-197 (2017-09-26)
Many infections and stress signals can rapidly activate the NLRP3 inflammasome to elicit robust inflammatory responses. This activation requires a priming step, which is thought to be mainly for upregulating NLRP3 transcription. However, recent studies report that the NLRP3 inflammasome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service