Skip to Content
Merck
All Photos(1)

Key Documents

11317

SAFC

Methanesulfonic acid

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3H
CAS Number:
Molecular Weight:
96.11
Beilstein:
1446024
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

biological source

synthetic

Quality Level

vapor density

3.3 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

98.0-102.0% (1 M NaOH, T)

form

liquid

impurities

≤5 ppm ethyl methansesulfonate (GC)
≤5 ppm isopropyl methanesulfonate (GC)
≤5 ppm methanesulfonyl chloride (GC)
≤5 ppm methyl methanesulfonate (GC)

evapn. residue

≤0.1%

refractive index

n20/D 1.429 (lit.)

bp

167 °C/10 mmHg (lit.)

mp

17-19 °C (lit.)

density

1.475-1.485 g/mL at 20 °C
1.481 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 ppm
sulfate (SO42-): ≤200 ppm

cation traces

Ta: ≤5 ppm

suitability

corresponds for IR spectroscopy

SMILES string

O=S(O)(C)=O

InChI

1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)

InChI key

AFVFQIVMOAPDHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

372.2 °F - closed cup

Flash Point(C)

189 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chris Twelves et al.
Breast cancer research and treatment, 148(3), 553-561 (2014-11-09)
Data from two phase 3 studies of eribulin were pooled in analyses initially requested by the European Medicines Agency to assess whether specific patient subgroups, previously treated with an anthracycline and a taxane, benefited from eribulin. Study 305/EMBRACE included women
Eric D Nacsa et al.
Organic letters, 15(1), 38-41 (2012-12-15)
The cyclopropenone catalyzed nucleophilic substitution of alcohols by methanesulfonate ion with inversion of configuration is described. This work provides an alternative to the Mitsunobu reaction that avoids the use of azodicarboxylates and generation of hydrazine and phosphine oxide byproducts. This
Kay O Johswich et al.
Infection and immunity, 83(1), 354-363 (2014-11-05)
Neisseria meningitidis asymptomatically colonizes the human upper respiratory tract but is also the cause of meningitis and severe septicemia. Carriage or disease evokes an immune response against the infecting strain. Hitherto, we have known little about the breadth of immunity
Renata Solarska et al.
Nanoscale, 4(5), 1553-1556 (2012-02-01)
Nanostructuring of semiconductor films offers the potential means for producing photoelectrodes with improved minority charge carrier collection. Crucial to the effective operation of the photoelectrode is also the choice of a suitable electrolyte. The behaviour of the nanostructured WO(3) photoanodes
Solvejg Jørgensen et al.
Physical chemistry chemical physics : PCCP, 15(14), 5140-5150 (2013-03-02)
The gas phase reaction between methane sulfonic acid (CH3SO3H; MSA) and the hydroxyl radical (HO), without and with a water molecule, was investigated with DFT-B3LYP and CCSD(T)-F12 methods. For the bare reaction we have found two reaction mechanisms, involving proton

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service