Skip to Content
Merck
All Photos(1)

Key Documents

08416

Sigma-Aldrich

(S)-4-Benzyloxazolidine-2-thione

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NOS
CAS Number:
Molecular Weight:
193.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0%

optical purity

enantiomeric excess: ≥98.0% (HPLC)

mp

63-67 °C

functional group

phenyl

SMILES string

S=C1N[C@H](CO1)Cc2ccccc2

InChI

1S/C10H11NOS/c13-10-11-9(7-12-10)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,11,13)/t9-/m0/s1

InChI key

WJSUXYCBZFLXIK-VIFPVBQESA-N

Application

A highly selective and efficient chiral auxiliary[1] which can be directly reduced to its corresponding aldehyde and the chiral auxiliary by reductive cleavage with diisobutylaluminum hydride.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Velazquez. F.; Olivo, H. F.
Current Organic Chemistry, 6, 303-303 (2002)
M T Crimmins et al.
Organic letters, 2(6), 775-777 (2001-02-07)
[formula: see text] Asymmetric aldol additions using chlorotitanium enolates of thiazolidinethione propionates proceed with high diastereoselectivity for the "Evans" or "non-Evans" syn product depending on the nature and amount of the base used. With (-)-sparteine as the base, selectivities of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service