Skip to Content
Merck
All Photos(1)

Key Documents

07817

Millipore

Oxidase Reagent according to Gaby-Hadley B

suitable for microbiology

Synonym(s):

Gaby-Hadley Reagent B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

Agency

according to ISO 9308-1:2014

Quality Level

product line

BioChemika

form

liquid

shelf life

limited shelf life, expiry date on the label

composition

N,N-dimethyl-p-phenylenediamine (oxalate), 0.1 g
dist. water, 100 mL

technique(s)

microbe id | specific enzyme detection: suitable

antibiotic activity spectrum

Gram-negative bacteria

application(s)

agriculture
clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

Plesiomonas spp.
Aeromonas spp.
Neisseria spp.
Pseudomonas spp.

General description

Oxidase Reagent can be used to detect oxidase positive organisms. The reagent is colorless in its reduced state and dark purple in its oxidized state. On applying to an oxidase-positive colony, it changes color.

Application

The reagent is used for detection of oxidase activity of a bacterial culture along with Oxidase Reagent A according to Gaby-Hadley.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neelima Garg
Laboratory Manual of Food Microbiology (2010)

Articles

Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

Specific agars and broths support detection and cultivation of Campylobacter, requiring complex media with essential supplements.

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

See All

Protocols

General guidance for the detection and enumeration of Enterobacteriaceae in food according to EN-ISO 8523:1991 and EN-ISO 4832:1991, respectively.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service