Skip to Content
Merck
All Photos(1)

Documents

07751

Millipore

Peptone, mycological No. 2

suitable for microbiology

Synonym(s):

Fungal peptone No. 2, Mycological Peptone No. 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.85

biological source

Porcine pancreas
Porcine skin

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥1.5%
total nitrogen (N), ≥12%

packaging

pkg of 500 g

storage condition

(Tightly closed. Dry.)

ign. residue

≤15%

loss

≤8.0% loss on drying

pH

5.0-6.0 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear (light yellow and light brown-yellow and light brown)

application(s)

microbiology

General description

Peptone, mycological No. 2, is a mixture of Porcine skin and pancreas-based peptones used for obtaining luxuriant growth of yeast and molds. It is a type of peptone that is widely used as a nutrient source in microbiological culture media for the cultivation of pathogenic and nonpathogenic fungi and the identification of saprophytic and dermatophytes -yeasts and molds.Peptone, mycological No. 2, provides a wide range of nutrients, including amino acids, vitamins, minerals, and peptides, which supports the growth of diverse fungi

Application

Peptone, mycological No. 2, is commonly used as an ingredient in many microbial culture media for the growth and identification of yeast and molds from various pharmaceutical, environmental, and food and beverage samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service