Skip to Content
Merck
All Photos(1)

Documents

06246

Supelco

Clenbuterol-d9

analytical standard

Synonym(s):

1-(4-Amino-3,5-dichlorophenyl)-2-(tert-butyl-d9-amino)ethanol, 4-Amino-α-(tert-butyl-d9-aminomethyl)-3,5-dichlorobenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D9H9Cl2N2O
CAS Number:
Molecular Weight:
286.25
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥97.0% (isotopic purity)
≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

environmental

mass shift

M+9

SMILES string

[2H]C([2H])([2H])C(NCC(O)c1cc(Cl)c(N)c(Cl)c1)(C([2H])([2H])[2H])C([2H])([2H])[2H]

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706019, T706030, T706020

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service