Skip to Content
Merck
All Photos(1)

Key Documents

05576

Supelco

Boron trifluoride-ethanol

~10% in ethanol (~1.3 M), LiChropur, for GC derivatization

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.22

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

concentration

~10% in ethanol (~1.3 M)

technique(s)

gas chromatography (GC): suitable

density

0.852 g/mL at 20 °C

storage temp.

2-8°C

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service