Skip to Content
Merck
All Photos(1)

Key Documents

05135001

Silybum marianum extract

secondary reference standard

Synonym(s):

Milk thistle extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

matrix material

storage temp.

−20°C

InChI

1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-16-5-3-12(7-18(16)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3/t20-,23?,24+,25-/m0/s1

InChI key

SEBFKMXJBCUCAI-VGHNRKBZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.
Silybum marianum (L.) Gaertner, fructus extract (according to HMPC Monograph)

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Silybin A+B (quantitative); Silicristin, Silidianin, Isosilibinin A, Isosilibinin B (qualitative)

Matrix Group: Botanicals

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service