Skip to Content
Merck
All Photos(1)

Documents

04423

Sigma-Aldrich

Phosphinic acid

purum, ≥60%

Synonym(s):

Hypophosphorous acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO2
CAS Number:
Molecular Weight:
66.00
EC Number:
MDL number:
UNSPSC Code:
12352106

grade

purum

concentration

≥60%

impurities

≤0.002% heavy metals (as Pb)

density

1.206 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

As: ≤2 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
Na: ≤1000 mg/kg

SMILES string

O[PH2]=O

InChI

1S/H3O2P/c1-3-2/h3H2,(H,1,2)

InChI key

ACVYVLVWPXVTIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service