Skip to Content
Merck
All Photos(1)

Key Documents

37-0745

Sigma-Aldrich

Zirconium(IV) butoxide solution

CP, 90% in 1-butanol

Synonym(s):

Tetrabutyl zirconate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(OC4H9)4
CAS Number:
Molecular Weight:
383.68
Beilstein:
4132649
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

CP

reaction suitability

reagent type: catalyst
core: zirconium

availability

available only in Japan

concentration

90% in 1-butanol

SMILES string

CCCCO[Zr](OCCCC)(OCCCC)OCCCC

InChI

1S/4C4H9O.Zr/c4*1-2-3-4-5;/h4*2-4H2,1H3;/q4*-1;+4

InChI key

BSDOQSMQCZQLDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in an unconventional method to prepare a gel of CeO2-stabilized zirconia polycrystals that have promising applications as protective coatings of steel and ferrous alloys.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service