Skip to Content
Merck
All Photos(1)

Key Documents

35-0380

Sigma-Aldrich

D-(+)-Xylose

SAJ special grade, 98.0-101.0%

Synonym(s):

D-xylopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1562108
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ special grade

Assay

98.0-101.0%

form

solid

availability

available only in Japan

mp

154-158 °C (lit.)

SMILES string

O[C@@H]1COC(O)[C@H](O)[C@H]1O

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3+,4-,5?/m1/s1

InChI key

SRBFZHDQGSBBOR-IOVATXLUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kilaru et al.
Fungal genetics and biology : FG & B, 79, 166-173 (2015-06-21)
Development of new fungicides, needed for sustainable control of fungal plant pathogens, requires identification of novel anti-fungal targets. Essential fungal-specific proteins are good candidates, but due to their importance, gene deletion mutants are not viable. Consequently, their cellular role often
Thu V Vuong et al.
PloS one, 9(4), e95170-e95170 (2014-04-17)
The xylan-binding module Clostridium thermocellum CBM22A was successfully fused to a gluco-oligosaccharide oxidase, GOOX-VN, from Sarocladium strictum via a short TP linker, allowing the fused protein to effectively bind different xylans. The presence of the CtCBM22A at the N-terminal of
Heather A Feaga et al.
mBio, 5(6), e01916-e01916 (2014-11-13)
Bacterial ribosomes frequently translate to the 3' end of an mRNA without terminating at a stop codon. Almost all bacteria use the transfer-messenger RNA (tmRNA)-based trans-translation pathway to release these "nonstop" ribosomes and maintain protein synthesis capacity. trans-translation is essential
Jose Serate et al.
Biotechnology for biofuels, 8, 180-180 (2015-11-20)
Microbial conversion of lignocellulosic feedstocks into biofuels remains an attractive means to produce sustainable energy. It is essential to produce lignocellulosic hydrolysates in a consistent manner in order to study microbial performance in different feedstock hydrolysates. Because of the potential
Wellington Muchero et al.
BMC genomics, 16, 24-24 (2015-01-24)
QTL cloning for the discovery of genes underlying polygenic traits has historically been cumbersome in long-lived perennial plants like Populus. Linkage disequilibrium-based association mapping has been proposed as a cloning tool, and recent advances in high-throughput genotyping and whole-genome resequencing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service