Skip to Content
Merck
All Photos(1)

Key Documents

28-6160

Sigma-Aldrich

Sulfuryl chloride

SAJ first grade, ≥90.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SO2Cl2
CAS Number:
Molecular Weight:
134.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.7 (vs air)

vapor pressure

100 mmHg ( 17.8 °C)
105 mmHg ( 20 °C)

Assay

≥90.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.443 (lit.)

bp

68-70 °C (lit.)

mp

−54 °C (lit.)

density

1.665 g/mL at 20 °C (lit.)

SMILES string

ClS(Cl)(=O)=O

InChI

1S/Cl2O2S/c1-5(2,3)4

InChI key

YBBRCQOCSYXUOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Harmata et al.
The Journal of organic chemistry, 72(2), 683-685 (2007-01-16)
Sulfinamides were synthesized from sulfonyl chlorides using a procedure involving in situ reduction of sulfonyl chlorides. The reaction is broad in scope and easy to perform.
Vlad Martin-Diaconescu et al.
Inorganic chemistry, 48(3), 1038-1044 (2009-01-10)
The electronic structure of organic sulfonyl compounds of the form RSO(2)G (G = -Cl, -OH, -CH(3)) is investigated to evaluate the effect of aryl R groups on photocleavage of the S-G bond. Sulfur K-edge X-ray absorption spectroscopy (XAS) provides a
Iron-catalyzed desulfinylative C-C cross-coupling reactions of sulfonyl chlorides with grignard reagents.
Chandra M Rao Volla et al.
Angewandte Chemie (International ed. in English), 47(7), 1305-1307 (2008-01-05)
Christopher Blackburn
ACS combinatorial science, 14(3), 150-154 (2012-02-11)
Reductive aminations and further transformations of an azo dye and fluorous tagged aldehyde are described. The intensely colored 2,4-dialkoxybenzyl protected amines undergo Fmoc-based peptide coupling, Suzuki reactions, and sulfonamide formation with product isolation facilitated by visual monitoring of fluorous solid
Zhiyong Wu et al.
Organic letters, 15(6), 1270-1273 (2013-03-07)
An efficient and concise one-pot protocol to synthesize sulfonylated quinoline N-oxides via copper-catalyzed C-H bond activation has been developed. Commercially available and less expensive aryl sulfonyl chlorides were used as the sulfonylation reagents. Various 2-aryl sulfonyl quinolines were obtained in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service