Skip to Content
Merck
All Photos(1)

Key Documents

28-6035

Sigma-Aldrich

Sulfuric acid (1+2)

~47.0 % (w/w) in H2O

Synonym(s):

Sulfuric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

reagent

form

liquid

availability

available only in Japan

concentration

~47.0 % (w/w) in H2O

density

1.37 g/cm3 at 20 °C

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young Hoon Jung et al.
Bioresource technology, 132, 109-114 (2013-02-12)
Dilute sulfuric acid pretreatment of oil palm empty fruit bunches (EFB) followed by the whole slurry fermentation of the pretreated EFB slurry was investigated. The optimized pretreatment conditions were at 1% (w/v) sulfuric acid with 3 min ramping to 190
Talita M Lacerda et al.
Carbohydrate polymers, 93(1), 347-356 (2013-03-08)
The influence of sulfuric acid concentration (H2SO4 5-25%, 100°C), crystallinity and fibers size on the hydrolysis reaction of sisal pulps were investigated, with the goal of evaluating both the liquor composition, as an important step in the production of bioethanol
Sanaa Pirani et al.
Carbohydrate polymers, 93(1), 357-363 (2013-03-08)
Cellulose consists of amorphous and crystalline regions. It is the crystalline regions which may be exploited to produce nanocrystalline cellulose (NCC). In order to extract nanocrystalline cellulose from native cellulose, sulfuric acid hydrolysis is typically used. The amorphous regions of
Volkmar Passoth et al.
Enzyme and microbial technology, 52(2), 105-110 (2013-01-01)
Integrated storage and pre-treatment (ISP) combines biopreservation of moist material under airtight conditions and pre-treatment. Moist wheat straw was inoculated with the biocontrol yeast Wickerhamomyces anomalus, the xylan degrading yeast Scheffersomyces stipitis or a co-culture of both. The samples and
M O Amdur
Environmental health perspectives, 81, 109-113 (1989-05-01)
Data from exposure of experimental animals and human subjects to sulfuric acid presents a consistent picture of its toxicology. Effects on airway resistance in asthmatic subjects were well predicted by data obtained on guinea pigs. Sulfuric acid increases the irritant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service