Skip to Content
Merck
All Photos(1)

Documents

28-3960

Sigma-Aldrich

Sodium pyrophosphate decahydrate

JIS special grade, ≥99.0%

Synonym(s):

Sodium pyrophosphate tetrabasic decahydrate, Tetrasodium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

9.5-10.5 (20 °C, 50 g/L)

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4Na.H4O7P2.10H2O/c;;;;1-8(2,3)7-9(4,5)6;;;;;;;;;;/h;;;;(H2,1,2,3)(H2,4,5,6);10*1H2/q4*+1;;;;;;;;;;;/p-4

InChI key

VZWGHDYJGOMEKT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruce L Riser et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 26(10), 3349-3357 (2011-03-15)
The high rate of cardiovascular mortality in patients with end-stage renal disease (ESRD) is a significant barrier to improved life expectancy. Unique in this population is the marked development and aggressive worsening of vascular calcification (VC). Pyrophosphate (PPi), an endogenous
Priyadip Das et al.
Chemical communications (Cambridge, England), 47(28), 8118-8120 (2011-06-21)
Newly synthesized Zn(II) and Cd(II)-based complexes show unique selectivity towards inorganic pyrophosphate (PPi) in 100% aqueous medium at pH = 7.4 and act as a "turn-on" and "turn-off" real-time assay, respectively, for the evaluation of the enzymatic activity of alkaline
Nadia Marino et al.
Inorganic chemistry, 50(1), 378-389 (2010-12-01)
The reaction in water of Cu(NO(3))(2)·2.5H(2)O with 2,2'-bipyridine (bipy), 1,10-phenanthroline (phen), or 1,10-phenanthroline-5-amine (phenam), and sodium pyrophosphate (Na(4)P(2)O(7)), at various pHs, afforded three new copper(II)-pyrophosphate complexes, namely, {[Cu(bipy)(cis-H(2)P(2)O(7))](2)}·3H(2)O (1a), {[Cu(phen)(H(2)O)](4)(HP(2)O(7))(2)}(ClO(4))(2)·4H(2)O (2), and {[Cu(2)(phenam)(2)(P(2)O(7))](2)·25H(2)O}(n) (3). A solvent free crystalline phase of
Taís Scaramucci et al.
Journal of dentistry, 39(12), 841-848 (2011-09-29)
To evaluate the erosive potential of orange juice modified by food additives in enamel and dentine. Calcium lactate pentahydrate (CLP), xanthan gum (XG), sodium linear polyphosphate (LPP), sodium pyrophosphate tetrabasic (PP), sodium tripolyphosphate (STP) and some of their combinations were
Catello Di Martino et al.
Planta, 234(4), 657-670 (2011-05-21)
Current studies in plants suggest that the content of the coenzyme NAD is variable and potentially important in determining cell fate. In cases that implicate NAD consumption, re-synthesis must occur to maintain dinucleotide pools. Despite information on the pathways involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service