Skip to Content
Merck
All Photos(1)

Documents

28-1590

Sigma-Aldrich

Sodium acetate

JIS special grade, ≥98.5%

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.5%

form

solid

autoignition temp.

1112 °F

availability

available only in Japan

pH

8.5-9.9 (25 °C, 246 g/L)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

storage temp.

15-25°C

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenta Iitani et al.
Analytical chemistry, 91(15), 9458-9465 (2019-07-10)
We developed a gas-imaging system (sniff-cam) for gaseous ethanol (EtOH) with improved sensitivity. The sniff-cam was applied to measure the extremely low concentration distribution of breath EtOH without the consumption of alcohol, which is related to the activity of the
Graham R Scott et al.
Molecular biology and evolution, 32(8), 1962-1976 (2015-04-09)
At high-altitude, small mammals are faced with the energetic challenge of sustaining thermogenesis and aerobic exercise in spite of the reduced O2 availability. Under conditions of hypoxic cold stress, metabolic demands of shivering thermogenesis and locomotion may require enhancements in
Mari Endo et al.
Journal of gastroenterology, 49(6), 1026-1039 (2013-07-13)
Daikenchuto (DKT), a gastrointestinal prokinetic Japanese herbal medicine, is prescribed for patients with postoperative ileus (POI) and adhesive bowel obstruction following abdominal surgery. Several mechanisms for the amelioration of POI by DKT have been suggested; however, it has remained unclear
Amanda P De Souza et al.
Journal of experimental botany, 66(14), 4351-4365 (2015-04-25)
The production of bioenergy from grasses has been developing quickly during the last decade, with Miscanthus being among the most important choices for production of bioethanol. However, one of the key barriers to producing bioethanol is the lack of information
Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service