Skip to Content
Merck
All Photos(1)

Key Documents

24-4260

Sigma-Aldrich

Potassium carbonate

SAJ first grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
grade:
SAJ first grade
form:
crystalline

grade

SAJ first grade

Assay

≥99.5%

form

crystalline

availability

available only in Japan

pH

11.0-13.0 (25 °C, 138 g/L)

mp

891 °C (lit.)

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Y Foo et al.
Bioresource technology, 102(20), 9814-9817 (2011-08-30)
Rice husk (RH), an abundant by-product of rice milling, was used for the preparation of activated carbon (RHAC) via KOH and K(2)CO(3) chemical activation. The activation process was performed at the microwave input power of 600 W for 7 min.
Carlos Arroniz et al.
Organic letters, 15(4), 910-913 (2013-02-05)
ortho-Arylation of ortho-substituted benzoic acids is a challenging process due to the tendency of the reaction products toward Pd-catalyzed protodecarboxylation. A simple method for preventing decarboxylation in sterically hindered benzoic acids is reported. The method described represents a reliable and
K Y Foo et al.
Bioresource technology, 104, 679-686 (2011-11-22)
This work explores the feasibility of orange peel, a citrus processing biomass as an alternative precursor for preparation of activated carbon (OPAC) via microwave assisted K(2)CO(3) activation. The operational parameters, chemical impregnation ratio, microwave power and irradiation time on the
K Y Foo et al.
Bioresource technology, 102(20), 9794-9799 (2011-08-31)
Sunflower seed oil residue, a by-product of sunflower seed oil refining, was utilized as a feedstock for preparation of activated carbon (SSHAC) via microwave induced K(2)CO(3) chemical activation. SSHAC was characterized by Fourier transform infrared spectroscopy, nitrogen adsorption-desorption and elemental
Yusuke Yagi et al.
Organic & biomolecular chemistry, 13(4), 1113-1121 (2014-11-26)
Fluorine-18 labeled radiotracers, such as [(18)F]fluorodeoxyglucose, can be used as practical diagnostic agents in positron emission tomography (PET). Furthermore, the properties of pharmaceuticals can be enhanced significantly by the introduction of fluorine groups into their original structures, and significant progress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service