Skip to Content
Merck
All Photos(1)

Key Documents

23-0230

Sigma-Aldrich

Bergamot oil

CP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211

grade

CP

form

oil

availability

available only in Japan

refractive index

n20/D 1.4665 (lit.)
n20/D 1.4675 (lit.)

bp

159 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

So Young Chang
Taehan Kanho Hakhoe chi, 38(4), 493-502 (2008-08-30)
The purpose of this study was to examine the effects of aroma hand massage on pain, state anxiety and depression in hospice patients with terminal cancer. This study was a nonequivalent control group pretest-posttest design. The subjects were 58 hospice
Carlo Bicchi et al.
Journal of chromatography. A, 1217(7), 1106-1113 (2009-10-23)
Asymmetrically substituted 6(I-VII)-O-t-butyldimethylsilyl(TBDMS)-3(I-VII)-O-ethyl-2(I-VII)-O-methyl-beta-cyclodextrin (MeEt-CD) and 6(I-VII)-O-TBDMS-2(I-VII)-O-ethyl-3(I-VII)-O-methyl-beta-cyclodextrin (EtMe-CD) were synthesised to evaluate the role of the substitution pattern in positions 2 and 3 on the enantioselectivity, in particular in view of their application to routine analysis in fast enantioselective gas chromatography
Laura Berliocchi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(11), 2780-2792 (2011-09-01)
Cosmetic, pharmaceutical, food and confectionary industries make increasing use of plant extracts in their products. Despite the widespread use of products containing plant extracts, the mechanisms of their effects are not fully characterized. Bergamot essential oil (BEO; Citrus bergamia, Risso)
Jin-Hee Hwang
Taehan Kanho Hakhoe chi, 36(7), 1123-1134 (2007-01-11)
The purpose of this study was to identify the effects of aromatherapy on blood pressure and stress responses of clients with essential hypertension. There were fifty-two subjects divided into an essential oil group, placebo group, and control group by random
K Kejlová et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(7), 1298-1303 (2007-08-03)
The aim of this study was to clarify the differences in the phototoxicity of bergamot oil obtained from four different suppliers. Spectral and chemical analyses were performed to identify presence of photoactive compounds in the test samples. The phototoxicity was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service