Skip to Content
Merck
All Photos(1)

Documents

09-0770

Sigma-Aldrich

Ethyl alcohol, pure

≥99.5%, JIS special grade

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:

product name

Ethanol, JIS special grade, ≥99.5%

grade

JIS special grade

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F

availability

available only in Japan

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L)

bp

78 °C (lit.)

mp

−114 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Poly(sulfobetaine) versus poly(ethylene glycol) based copolymer modified polyurethane catheters for antifouling: This study examines the use of ethanol in the synthesis of polymers designed to reduce biological fouling on medical devices, highlighting its role in advanced healthcare materials research (Cao et al., 2024).

  • Green synthesis of polyimide by using an ethanol solvothermal method for aqueous zinc batteries: Ethanol is used as a green solvent in the innovative synthesis of polyimide structures, emphasizing its importance in sustainable material production and energy storage solutions (Zhao et al., 2024).

  • Modulation of gut microbiota with probiotics as a strategy to counteract endogenous and exogenous neurotoxicity: Research explores the indirect applications of ethanol in the solvent extraction processes used for studying neuroprotective strategies in medical research (Skalny et al., 2024).

  • Superselective Ablative Chemo-ethanol Embolization for Recurrent Single Hepatocellular Carcinoma: A Six-Month Outcome Analysis: This clinical study utilizes ethanol directly in therapeutic interventions, demonstrating its critical use in oncological treatments to improve patient outcomes (Lee et al., 2024).

  • Self-Assembly Reinforced Alginate Fibers for Enhanced Strength, Toughness, and Bone Regeneration: Ethanol is employed in the processing of novel biomaterials designed for medical applications, showcasing its utility in the development of regenerative medicine technologies (Cui et al., 2024).

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Di Girolamo et al.
Stem cells (Dayton, Ohio), 33(1), 157-169 (2014-06-27)
Stem cell (SC) division, deployment, and differentiation are processes that contribute to corneal epithelial renewal. Until now studying the destiny of these cells in a living mammal has not been possible. However, the advent of inducible multicolor genetic tagging and
Ioannis Lignos et al.
Small (Weinheim an der Bergstrasse, Germany), 11(32), 4009-4017 (2015-05-23)
The early-time kinetics (<1 s) of lead sulfide (PbS) quantum dot formation are probed using a novel droplet-based microfluidic platform, which allows for high-throughput and real-time optical analysis of the reactive process with millisecond time resolution. The reaction platform enables
Ross B Mounsey et al.
Experimental neurology, 273, 36-44 (2015-08-06)
Parkinson's disease (PD) is a common chronic neurodegenerative disorder, usually of idiopathic origin. Symptoms including tremor, bradykinesia, rigidity and postural instability are caused by the progressive loss of dopaminergic neurons in the nigrostriatal region of the brain. Symptomatic therapies are
Rubaiyea Farrukee et al.
The Journal of antimicrobial chemotherapy, 70(7), 2004-2012 (2015-03-20)
The burden of disease due to influenza B is often underestimated. Clinical studies have shown that oseltamivir, a widely used neuraminidase inhibitor (NAI) antiviral drug, may have reduced effectiveness against influenza B viruses. Therefore, it is important to study the
Cem Kuscu et al.
Nature biotechnology, 32(7), 677-683 (2014-05-20)
RNA-guided genome editing with the CRISPR-Cas9 system has great potential for basic and clinical research, but the determinants of targeting specificity and the extent of off-target cleavage remain insufficiently understood. Using chromatin immunoprecipitation and high-throughput sequencing (ChIP-seq), we mapped genome-wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service