Skip to Content
Merck
All Photos(1)

Key Documents

05-0470

Sigma-Aldrich

Calcium carbide

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaC2
CAS Number:
Molecular Weight:
64.10
Beilstein:
3909011
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

CP

form

crystalline

availability

available only in Japan

pH

12.48 (20 g/L)

density

2.22 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[C-]#[C-]

InChI

1S/C2.Ca/c1-2;/q-2;+2

InChI key

UIXRSLJINYRGFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoguang Jiao et al.
Science in China. Series C, Life sciences, 48 Suppl 1, 26-32 (2005-08-11)
A field experiment was carried out at the Shenyang Experimental Station of Ecology (CAS) in order to study the effects of slow-release urea fertilizers high polymer-coated urea (SRU1), SRU1 mixed with dicyandiamide DCD (SRU2), and SRU1 mixed with calcium carbide
G Malla et al.
Chemosphere, 58(2), 141-147 (2004-12-02)
Mitigation of methane (CH4) and nitrous oxide (N2O) emissions from soil is important to reduce the global warming. Efficacy of five nitrification inhibitors, i.e. neem (Azadirachta melia) cake, thiosulphate, coated calcium carbide, neem oil coated urea and dicyandiamide (DCD) and
Production of calcium carbide from fine biochars.
Guodong Li et al.
Angewandte Chemie (International ed. in English), 49(45), 8480-8483 (2010-09-14)
H Kjuus et al.
British journal of industrial medicine, 43(4), 237-242 (1986-04-01)
The overall mortality and the incidence of cancer have been studied among male employees at a plant producing calcium carbide. The cohort was defined as all men employed at the plant for at least 18 months in the period 1953
P Boffetta et al.
Cancer causes & control : CCC, 8(3), 444-472 (1997-05-01)
Epidemiologic evidence on the relationship between polycyclic aromatic hydrocarbons (PAH) and cancer is reviewed. High occupational exposure to PAHs occurs in several industries and occupations. Covered here are aluminum production, coal gasification, coke production, iron and steel foundries, tar distillation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service