RIE156325145
Rieke Metals, Inc.
3'-Carboethoxy-2-morpholinomethyl benzophenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
ethyl 3-[2-(4-morpholinylmethyl)benzoyl]benzoate
Inchi Code
1S/C21H23NO4/c1-2-26-21(24)17-8-5-7-16(14-17)20(23)19-9-4-3-6-18(19)15-22-10-12-25-13-11-22/h3-9,14H,2,10-13,15H2,1H3
InChI key
GLMWMTYWULROBK-UHFFFAOYSA-N
Purity
97%
Country of Origin
US
Product Link
https://riekemetals.com/products-services/catnum/6126-0100-12
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service