RIE156320474
Rieke Metals, Inc.
2-ethoxycarbonyl-3'-iodobenzophenone
About This Item
Iupac Name
ethyl 2-(3-iodobenzoyl)benzoate
Inchi Code
1S/C16H13IO3/c1-2-20-16(19)14-9-4-3-8-13(14)15(18)11-6-5-7-12(17)10-11/h3-10H,2H2,1H3
InChI key
QGMNPDGUCSMKHT-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.