TANKT0PEQ
Milli-Q® EQ Storage Tank Top Assembly without ASM
Protects stored pure water from airborne contaminants
About This Item
Recommended Products
feature
Allows you to connect up to 3 water purification systems to one tank (25 L, 50 L or 100 L)
manufacturer/tradename
Milli-Q®
tank depth × H
26.0 cm (10.2 in.) × 10.0 cm (3.9 in.)
compatibility
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Tanks
shipped in
ambient
Related Categories
General description
For use with Milli-Q® EQ 7008/16 water purification systems.
Application
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service