S384PCR
MultiScreen PCR384 Filter Plate
Purification of PCR products
Synonym(s):
Montage PCR Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.15
Recommended Products
material
polystyrene
Quality Level
sterility
non-sterile
manufacturer/tradename
MultiScreen®
parameter
100 μL sample volume (per well)
technique(s)
PCR: suitable
plate size
384 wells
pore size
pore sizeproprietary
shipped in
ambient
Related Categories
General description
Filter Code: PCR
Application
Purification of PCR products
Research Category
Nucleic acid
Nucleic acid
Features and Benefits
Proprietary membrane
Components
Does not include reagents
Physical form
Color Code: Clear
Other Notes
Automation Compatible: No
Legal Information
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service