Skip to Content
Merck
All Photos(1)

Key Documents

PX0040-1

Sigma-Aldrich

Pancreatin Powder

Sign Into View Organizational & Contract Pricing

Select a Size

PX0040-1
€280.00

€280.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
PX0040-1
€280.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.77

€280.00


Estimated to ship on28 May 2025


Request a Bulk Order

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service